Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha M/CD11b, Clone: C11b/660, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene ID (Entrez) 3684
Format Purified
Cross Reactivity Human
Gene Accession No. P11215
Host Species Mouse
Immunogen Recombinant human CD11b protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Antigen Integrin alpha M/CD11b
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Clone C11b/660
Primary or Secondary Primary
Isotype IgG2b, κ
Concentration 1mg/mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Quantity 0.1mg