Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha L/CD11a, Clone: HI111, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen Integrin alpha L/CD11a
Clone HI111
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD11a.
Gene ID (Entrez) 3683
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344001

 
novus biologicals canada
NBP14344001MG
0.1mg Each for $285.00

NBP14344003

 
novus biologicals canada
NBP1434400025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen Integrin alpha L/CD11a
Clone HI111
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD11a.
Gene ID (Entrez) 3683
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)