Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha L/CD11a, Clone: HI111, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434400025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344003

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Antigen Integrin alpha L/CD11a
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was CD11a.
Purification Method Affinity Purified
Quantity 0.025mg
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3683
Clone HI111
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG1, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL