Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha L/CD11a, Clone: HI111, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344001

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Host Species Mouse
Antigen Integrin alpha L/CD11a
Quantity 0.1mg
Format Affinity Purified
Purification Method Affinity Purified
Conjugate Unlabeled
Clone HI111
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Primary or Secondary Primary
Gene ID (Entrez) 3683
Immunogen The immunogen for this antibody was CD11a.
Isotype IgG1, κ
Cross Reactivity Human
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.