Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha L/CD11a, Clone: CRIS-3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459901

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P20701
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3683
Reconstitution Protein A purified
Clone CRIS-3
Concentration 1mg/mL
Conjugate Unlabeled
Antigen Integrin alpha L/CD11a
Immunogen Stimulated human leukocytes
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.