missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha E/CD103, Clone: B-Ly7, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen Integrin alpha E/CD103
Clone B-Ly7
Immunogen The immunogen for this antibody was CD103/Integrin alpha E.
Gene ID (Entrez) 3682
Formulation Phosphate buffer pH 7.2
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14337003 Novus Biologicals Canada
NBP1433700025MG
0.025mg Each for $219.00
NBP14337001 Novus Biologicals Canada
NBP14337001MG
0.1mg Each for $285.00
Specifications Description & Specifications

Specifications

Integrin alpha E/CD103
The immunogen for this antibody was CD103/Integrin alpha E.
Phosphate buffer pH 7.2
Primary
Murine
Unlabeled
Affinity Purified
B-Ly7
3682
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen)