Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha E/CD103, Clone: B-Ly7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433700025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14337003

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Cross Reactivity Human
Immunogen The immunogen for this antibody was CD103/Integrin alpha E.
Quantity 0.025mg
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Antigen Integrin alpha E/CD103
Host Species Mouse
Purification Method Affinity Purified
Regulatory Status RUO
Gene ID (Entrez) 3682
Isotype IgG1, κ
Clone B-Ly7
Primary or Secondary Primary