Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha E/CD103, Clone: B-Ly7, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Integrin alpha E/CD103
Clone B-Ly7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD103/Integrin alpha E.
Gene ID (Entrez) 3682
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143979

 
novus biologicals canada
NBP143979100TESTS
100 Tests Each for $252.00

NBP14397925

 
novus biologicals canada
NBP14397925TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen Integrin alpha E/CD103
Clone B-Ly7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD103/Integrin alpha E.
Gene ID (Entrez) 3682
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry