Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha 5/CD49e, Clone: SAM-1, Biotin, Novus Biologicals™

$128.00 - $246.00

Specifications

Antigen Integrin alpha 5/CD49e
Clone SAM-1
Concentration 0.5mg/mL
Immunogen Native purified from CD49e-positive cells.
Gene ID (Entrez) 3678
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14363201

 
novus biologicals canada
NBP14363201MG
0.1mg Each for $246.00

NBP14363203

 
novus biologicals canada
NBP1436320025MG
0.025mg Each for $128.00
Description & Specifications

Specifications

Antigen Integrin alpha 5/CD49e
Clone SAM-1
Concentration 0.5mg/mL
Immunogen Native purified from CD49e-positive cells.
Gene ID (Entrez) 3678
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry