Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha 4/CD49d, Clone: 9F10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433980025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14339803

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Cross Reactivity Human
Antigen Integrin alpha 4/CD49d
Immunogen The immunogen for this antibody was CD49d.
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.025mg
Clone 9F10
Host Species Mouse
Conjugate Unlabeled
Isotype IgG1, κ
Purification Method Affinity Purified
Format Affinity Purified
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3676