Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM531, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Insulin
Clone SPM531
Concentration 0.2mg/mL
Immunogen Full length (1-84 amino acid) purified pig insulin
Gene Accession No. P01308
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23297501

 
novus biologicals canada
NBP23297501ML
0.1mL Each for $139.00

NBP23297510

 
novus biologicals canada
NBP23297510ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Insulin
Clone SPM531
Concentration 0.2mg/mL
Immunogen Full length (1-84 amino acid) purified pig insulin
Gene Accession No. P01308
Gene ID (Entrez) 3630
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting