Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM531, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297510

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Full length (1-84 amino acid) purified pig insulin
Format Purified
Gene Accession No. P01308
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Formulation PBS with 0.05% BSA.
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM531
Concentration 0.2mg/mL