Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM531, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Host Species Mouse
Quantity 0.1mg
Primary or Secondary Primary
Gene Accession No. P01308
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Clone SPM531
Antigen Insulin
Purification Method Purified
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Full length (1-84 amino acid) purified pig insulin
Concentration 1mg/mL
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 3630
Test Specificity Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide.