Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM139, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297410

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Antigen Insulin
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01308
Host Species Mouse
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Test Specificity Does not react to Mouse and Rat.
Reconstitution Protein A purified
Clone SPM139
Concentration 0.2mg/mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine