Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM139, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297401

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Immunogen Full length (1-84 amino acid) purified pig insulin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 3630
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Host Species Mouse
Format Purified
Antigen Insulin
Primary or Secondary Primary
Test Specificity Does not react to Mouse and Rat.
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Concentration 0.2mg/mL
Gene Accession No. P01308
Purification Method Purified
Clone SPM139
Reconstitution Protein A purified