Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: SPM139, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473801

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin
Gene Accession No. P01308
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Test Specificity Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Does not react with Mouse or Rat.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Reconstitution Protein A purified
Clone SPM139
Concentration 1mg/mL
Conjugate Unlabeled