Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23426005ML

 View all options for this product


For Research Use Only

Catalog No. NBP23426005


Description & Specifications


Antigen Insulin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone E2-E3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01308
Host Species Mouse
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Reconstitution Protein A purified