Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426001

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation PBS with 0.05% BSA.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Full length (1-84 amino acid) purified pig insulin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3630
Regulatory Status RUO
Host Species Mouse
Purification Method Purified
Isotype IgG1, κ
Clone E2-E3
Concentration 0.2mg/mL
Reconstitution Protein A purified
Quantity 0.1mL
Gene Accession No. P01308
Primary or Secondary Primary