Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Immunogen Full length (1-84 amino acid) purified pig insulin
Gene ID (Entrez) 3630
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Primary or Secondary Primary
Regulatory Status RUO
Concentration 1mg/mL
Format Purified
Reconstitution Protein A purified
Quantity 0.1mg
Isotype IgG1, κ
Antigen Insulin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01308
Purification Method Purified
Clone E2-E3