Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3+2D11-H5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426105

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen Insulin
Host Species Mouse
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01308
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Reconstitution Protein A purified
Clone E2-E3+2D11-H5
Concentration 0.2mg/mL