Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3+2D11-H5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Regulatory Status RUO
Format Purified
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Purification Method Purified
Concentration 1mg/mL
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Primary or Secondary Primary
Antigen Insulin
Quantity 0.1mg
Clone E2-E3+2D11-H5
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene ID (Entrez) 3630
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P01308