Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: E2-E3+2D11-H5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461202

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone E2-E3+2D11-H5
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01308
Host Species Murine
Immunogen Full length (1-84 amino acid) purified pig insulin
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Reconstitution Protein A purified
Target Species Bovine
Target Species Human
Target Species Porcine
Target Species Rabbit
Target Species Rat