Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: 2D11-H5, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen Insulin
Clone 2D11-H5
Concentration 0.2mg/mL
Immunogen Human insulin conjugated to BSA was used as immunogen to generate the insulin antibody.
Gene Accession No. P01308
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21519501

 
novus biologicals canada
NBP21519501MG
0.1mg Each for $329.00

NBP21519502

 
novus biologicals canada
NBP21519502MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen Insulin
Clone 2D11-H5
Concentration 0.2mg/mL
Immunogen Human insulin conjugated to BSA was used as immunogen to generate the insulin antibody.
Gene Accession No. P01308
Gene ID (Entrez) 3630
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot