Please login to your online account to display your discounted pricing

Mouse anti-Insulin, Clone: 2D11-H5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519502

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone 2D11-H5
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P01308
Host Species Murine
Immunogen Human insulin conjugated to BSA was used as immunogen to generate the insulin antibody.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3630
Test Specificity Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Target Species Bovine
Target Species Human
Target Species Porcine