Please login to your online account to display your discounted pricing

Mouse anti-ILT2/CD85j/LILRB1, Clone: HP-F1, Novus Biologicals™

$246.00 - $321.00

Specifications

Antigen ILT2/CD85j/LILRB1
Clone HP-F1
Concentration 1mg/mL
Immunogen The immunogen for this antibody was CD85j.
Gene ID (Entrez) 10859
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142998

 
novus biologicals canada
NBP142998100TESTS
100 Tests Each for $321.00

NBP14299825

 
novus biologicals canada
NBP14299825TESTS
25 Tests Each for $246.00
Description & Specifications

Specifications

Antigen ILT2/CD85j/LILRB1
Clone HP-F1
Concentration 1mg/mL
Immunogen The immunogen for this antibody was CD85j.
Gene ID (Entrez) 10859
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay