Please login to your online account to display your discounted pricing

Mouse anti-ILT2/CD85j/LILRB1, Clone: HP-F1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14299825TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14299825

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Cross Reactivity Human
Primary or Secondary Primary
Antigen ILT2/CD85j/LILRB1
Format Affinity Purified
Gene ID (Entrez) 10859
Host Species Mouse
Formulation Aqueous buffer (contains stabilizer if necessary)
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Concentration 1mg/mL
Clone HP-F1
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CD85j.
Quantity 25 Tests
Monoclonal or Polyclonal Monoclonal