Please login to your online account to display your discounted pricing

Mouse anti-ILT2/CD85j/LILRB1, Clone: HP-F1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP142998100TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP142998


Description & Specifications


Antigen ILT2/CD85j/LILRB1
Applications Flow Cytometry
Applications Functional Assay
Clone HP-F1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Mouse
Immunogen The immunogen for this antibody was CD85j.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10859