Please login to your online account to display your discounted pricing

Mouse anti-ILT2/CD85j/LILRB1, Clone: HP-F1, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen ILT2/CD85j/LILRB1
Clone HP-F1
Immunogen The immunogen for this antibody was CD85j.
Gene ID (Entrez) 10859
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14355401

 
novus biologicals canada
NBP14355401MG
0.1mg Each for $329.00

NBP14355403

 
novus biologicals canada
NBP1435540025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen ILT2/CD85j/LILRB1
Clone HP-F1
Immunogen The immunogen for this antibody was CD85j.
Gene ID (Entrez) 10859
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry