Please login to your online account to display your discounted pricing

IL-7 R alpha/CD127 Mouse anti-Human, Clone: 40131, R&D Systems

Mouse Monoclonal Antibody

$343.25 - $891.94

Specifications

Antigen IL-7 R alpha/CD127
Immunogen Extracellular domain
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MAB306500

 
r & d systems
MAB306500
500μg Each for $891.94

MAB306100

 
r & d systems
MAB306100
100μg Each for $343.25
Description & Specifications

Specifications

Antigen IL-7 R alpha/CD127
Immunogen Extracellular domain
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Ascites
Format Purified
Applications Flow Cytometry
Applications Western Blot
Regulatory Status RUO
Storage Requirements 2° to 8°C under sterile conditions after reconstitution; 6 months, −20° to −70°C under sterile conditions. Avoid repeated freeze-thaw cycles.

IL-7 R alpha/CD127 Monoclonal antibody MAB306 Specifically detects IL-7 R alpha/CD127 in Human samples. It is validated for Flow Cytometry, Western Blotting