Please login to your online account to display your discounted pricing

Mouse anti-IL-4, Clone: 8D4-8, PE, Novus Biologicals™

$128.00 - $289.00

Specifications

Antigen IL-4
Clone 8D4-8
Concentration 0.2mg/mL
Immunogen This antibody was generated by immunizing host with Yeast-expressed
Gene ID (Entrez) 3565
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143726

 
novus biologicals canada
NBP143726100TESTS
100 Tests Each for $289.00

NBP14372625

 
novus biologicals canada
NBP14372625TESTS
25 Tests Each for $128.00
Description & Specifications

Specifications

Antigen IL-4
Clone 8D4-8
Concentration 0.2mg/mL
Immunogen This antibody was generated by immunizing host with Yeast-expressed
Gene ID (Entrez) 3565
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry