Please login to your online account to display your discounted pricing

Mouse anti-IL-4, Clone: 8D4-8, APC, Novus Biologicals™

$246.00 - $354.00

Specifications

Antigen IL-4
Clone 8D4-8
Immunogen This antibody was generated by immunizing host with Yeast-expressed
Gene ID (Entrez) 3565
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14298305

 
novus biologicals canada
NBP142983005MG
0.05mg Each for $246.00

NBP14298301

 
novus biologicals canada
NBP14298301MG
0.1mg Each for $354.00
Description & Specifications

Specifications

Antigen IL-4
Clone 8D4-8
Immunogen This antibody was generated by immunizing host with Yeast-expressed
Gene ID (Entrez) 3565
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry