Please login to your online account to display your discounted pricing

Mouse anti-IL-4, Clone: 144BS, Novus Biologicals™

$384.00 - $934.00

Specifications

Antigen IL-4
Clone 144BS
Immunogen -C
Gene Accession No. Q8V42
Gene ID (Entrez) 3565
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP22517501

 
novus biologicals canada
NBP22517501MG
0.1mg Each for $384.00

NBP2251755

 
novus biologicals canada
NBP22517505MG
0.5mg Each for $934.00
Description & Specifications

Specifications

Antigen IL-4
Clone 144BS
Immunogen -C
Gene Accession No. Q8V42
Gene ID (Entrez) 3565
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunoprecipitation