Please login to your online account to display your discounted pricing

Mouse anti-IL-4, Clone: 144BS, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP22517501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP22517501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunoprecipitation
Formulation PBS. Protein G purified.
Quantity 0.1mg
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. Q8V42
Purification Method Purified
Regulatory Status RUO
Immunogen -C
Host Species Mouse
Format Purified
Monoclonal or Polyclonal Monoclonal
Isotype IgG
Clone 144BS
Gene ID (Entrez) 3565
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Antigen IL-4