Please login to your online account to display your discounted pricing

Mouse anti-IL-4, Clone: 144BS, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP22517505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2251755

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen IL-4
Format Purified
Formulation PBS. Protein G purified.
Gene Accession No. Q8V42
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunoprecipitation
Immunogen -C
Quantity 0.5mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 144BS
Conjugate Unlabeled
Isotype IgG
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 3565