Please login to your online account to display your discounted pricing

Mouse anti-IL-32, Clone: 2F9, Novus Biologicals™

$254.67 - $310.66

Specifications

Antigen IL-32
Clone 2F9
Concentration 1.0mg/mL
Immunogen Recombinant human IL32 (1-131aa) purified from E. coli
Gene Accession No. NP_001012651
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14528501

 
novus biologicals canada
NBP14528501ML
0.1mL Each for $310.66

NBP14528505

 
novus biologicals canada
NBP145285005ML
0.05mL Each for $254.67
Description & Specifications

Specifications

Antigen IL-32
Clone 2F9
Concentration 1.0mg/mL
Immunogen Recombinant human IL32 (1-131aa) purified from E. coli
Gene Accession No. NP_001012651
Gene ID (Entrez) 9235
Formulation In Phosphate-Buffered Saline (pH 7.4)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Western Blotting