Please login to your online account to display your discounted pricing

Mouse anti-IL-32, Clone: 2F9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP145285005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14528505

  • / Each
 

Description & Specifications

Specifications

Antigen IL-32
Applications ELISA
Applications Western Blot
Clone 2F9
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation In Phosphate-Buffered Saline (pH 7.4)
Gene Accession No. NP_001012651
Host Species Murine
Immunogen Recombinant human IL32 (1-131aa) purified from E. coli
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.05mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9235
Test Specificity This antibody is specific to Human IL32
Target Species Human