Please login to your online account to display your discounted pricing

Mouse anti-IL-3 R alpha/CD123, Clone: 6H6, PE-Cy7, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen IL-3 R alpha/CD123
Clone 6H6
Immunogen Tissue / cell preparation. IL3R transfected COS cells.
Gene ID (Entrez) 3563
Formulation aqueous bufffer may contain carrier protein/stabilizer.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200467

 
novus biologicals canada
NBP200467100TESTS
100 Tests Each for $362.00

NBP20046725

 
novus biologicals canada
NBP20046725TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen IL-3 R alpha/CD123
Clone 6H6
Immunogen Tissue / cell preparation. IL3R transfected COS cells.
Gene ID (Entrez) 3563
Formulation aqueous bufffer may contain carrier protein/stabilizer.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry