Please login to your online account to display your discounted pricing

Mouse anti-IL-3 R alpha/CD123, Clone: 6H6, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen IL-3 R alpha/CD123
Clone 6H6
Immunogen Tissue / cell preparation. IL3R transfected COS cells.
Gene ID (Entrez) 3563
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200468

 
novus biologicals canada
NBP200468100TESTS
100 Tests Each for $395.00

NBP20046825

 
novus biologicals canada
NBP20046825TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen IL-3 R alpha/CD123
Clone 6H6
Immunogen Tissue / cell preparation. IL3R transfected COS cells.
Gene ID (Entrez) 3563
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry