Please login to your online account to display your discounted pricing

Mouse anti-IL-23A/IL-23 P19, Clone: 473P19, Novus Biologicals™

$274.00 - $648.00

Specifications

Antigen IL-23A/IL-23 P19
Clone 473P19
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL23 p19.
Gene Accession No. Q9NPF7
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14323703

 
novus biologicals canada
NBP1432370025MG
0.025mg Each for $274.00

NBP1432375

 
novus biologicals canada
NBP14323705MG
0.5mg Each for $648.00
Description & Specifications

Specifications

Antigen IL-23A/IL-23 P19
Clone 473P19
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL23 p19.
Gene Accession No. Q9NPF7
Gene ID (Entrez) 51561
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Western Blotting