Please login to your online account to display your discounted pricing

Mouse anti-IL-23A/IL-23 P19, Clone: 473P19, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14323705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432375

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Cross Reactivity Human
Gene ID (Entrez) 51561
Format Affinity Purified
Primary or Secondary Primary
Host Species Mouse
Immunogen The immunogen for this antibody was IL23 p19.
Quantity 0.5mg
Conjugate Unlabeled
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Antigen IL-23A/IL-23 P19
Isotype IgG1, κ
Concentration 0.5mg/mL
Formulation Phosphate buffer pH 7.2
Clone 473P19
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Gene Accession No. Q9NPF7