Please login to your online account to display your discounted pricing

Mouse anti-IL-23A/IL-23 P19, Clone: 473P19, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432370025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14323703

  • / Each
 

Description & Specifications

Specifications

Antigen IL-23A/IL-23 P19
Host Species Mouse
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Format Affinity Purified
Gene Accession No. Q9NPF7
Applications ELISA
Applications Western Blotting
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 51561
Clone 473P19
Concentration 0.5mg/mL
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was IL23 p19.
Conjugate Unlabeled