Please login to your online account to display your discounted pricing

Mouse anti-IL-17F, Clone: H17F10A7, Novus Biologicals™

$307.00 - $406.00

Specifications

Antigen IL-17F
Clone H17F10A7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL17F.
Gene ID (Entrez) 112744
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14323401

 
novus biologicals canada
NBP14323401MG
0.1mg Each for $406.00

NBP14323403

 
novus biologicals canada
NBP1432340025MG
0.025mg Each for $307.00
Description & Specifications

Specifications

Antigen IL-17F
Clone H17F10A7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL17F.
Gene ID (Entrez) 112744
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blotting