Please login to your online account to display your discounted pricing

Mouse anti-IL-17F, Clone: H17F10A7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432340025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14323403

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Antigen IL-17F
Regulatory Status RUO
Primary or Secondary Primary
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.025mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 112744
Clone H17F10A7
Conjugate Unlabeled
Immunogen The immunogen for this antibody was IL17F.
Isotype IgG1
Concentration 0.5mg/mL
Cross Reactivity Human