Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: CRM57, Biotin, Novus Biologicals™

$197.00 - $549.00

Specifications

Antigen IL-1 beta/IL-1F2
Clone CRM57
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL1b.
Gene ID (Entrez) 3553
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1434935

 
novus biologicals canada
NBP14349305MG
0.5mg Each for $549.00

NBP1434935M

 
novus biologicals canada
NBP143493005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen IL-1 beta/IL-1F2
Clone CRM57
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL1b.
Gene ID (Entrez) 3553
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA