Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: CRM56, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143245005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14324505

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Neutralization
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody was IL1b.
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone CRM56
Conjugate Unlabeled
Cross Reactivity Human
Antigen IL-1 beta/IL-1F2
Formulation Phosphate buffer pH 7.2
Isotype IgG1
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3553