Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: CRM56, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14324505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432455

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Neutralization
Conjugate Unlabeled
Quantity 0.5mg
Immunogen The immunogen for this antibody was IL1b.
Monoclonal or Polyclonal Monoclonal
Antigen IL-1 beta/IL-1F2
Format Affinity Purified
Host Species Mouse
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Isotype IgG1
Gene ID (Entrez) 3553
Clone CRM56
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary