Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: CRM56, FITC, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen IL-1 beta/IL-1F2
Clone CRM56
Immunogen The immunogen for this antibody was IL1b.
Gene ID (Entrez) 3553
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143929

 
novus biologicals canada
NBP143929100TESTS
100 Tests Each for $296.00

NBP14392925

 
novus biologicals canada
NBP14392925TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen IL-1 beta/IL-1F2
Clone CRM56
Immunogen The immunogen for this antibody was IL1b.
Gene ID (Entrez) 3553
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry