Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: 1400.24.17, Novus Biologicals™

$384.00 - $1,077.00

Specifications

Antigen IL-1 beta/IL-1F2
Clone 1400.24.17
Concentration 1.0mg/mL
Immunogen This antibody recognizes both the recombinant and the natural form of mouse IL-1b.
Gene ID (Entrez) 3553
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23115401

 
novus biologicals canada
NBP23115401MG
0.1mg Each for $384.00

NBP2311545

 
novus biologicals canada
NBP23115405MG
0.5mg Each for $1,077.00
Description & Specifications

Specifications

Antigen IL-1 beta/IL-1F2
Clone 1400.24.17
Concentration 1.0mg/mL
Immunogen This antibody recognizes both the recombinant and the natural form of mouse IL-1b.
Gene ID (Entrez) 3553
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Mouse
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Blocking
Applications ELISA
Applications Neutralization