Please login to your online account to display your discounted pricing

Mouse anti-IL-1 beta/IL-1F2, Clone: 1400.24.17, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23115401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23115401

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications ELISA
Applications Neutralization
Cross Reactivity Mouse
Quantity 0.1mg
Immunogen This antibody recognizes both the recombinant and the natural form of mouse IL-1b.
Formulation PBS. Protein G purified.
Gene ID (Entrez) 3553
Test Specificity 10pg/mL to 3ng/mL
Clone 1400.24.17
Purification Method Purified
Concentration 1.0mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Antigen IL-1 beta/IL-1F2
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Host Species Mouse