Please login to your online account to display your discounted pricing

Mouse anti-IgM, Clone: SPM557, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Antigen IgM
Host Species Mouse
Immunogen Recombinant heavy chain of human IgM
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3507
Clone SPM557
Concentration 0.2mg/mL
Conjugate Unlabeled
Test Specificity Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Reconstitution Protein A purified
Cross Reactivity Human